Anti-TNFRSF14 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 94801]

Supplier: R&D Systems
FAB356A
220017-625EA 605.94 USD
220017-625
Anti-TNFRSF14 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 94801]
Antibodies
Type: Primary
Antigen: TNFRSF14
Clonality: Monoclonal
Clone: 94801
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR