Anti-SDC3 Goat Polyclonal Antibody (APC (Allophycocyanin))

Supplier: R&D Systems
FAB3539A
220017-621EA 634.86 USD
220017-621
Anti-SDC3 Goat Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Type: Primary
Antigen: SDC3
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR