Anti-FLT4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 54733]

Supplier: R&D Systems
FAB3492P
220017-616EA 591.48 USD
220017-616
Anti-FLT4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 54733]
Antibodies
Type: Primary
Antigen Symbol: FLT4
Antigen Name: fms-related tyrosine kinase 4
Antigen Synonyms: FLT41, LMPH1A, PCL, VEGFR3
Clonality: Monoclonal
Clone: 54733
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR