Anti-EPHB4 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 395810]

Supplier: R&D Systems
FAB3038P
220017-538EA 548.09 USD
220017-538
Anti-EPHB4 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 395810]
Antibodies
Type: Primary
Antigen: EPHB4
Clonality: Monoclonal
Clone: 395810
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR