Anti-CEACAM1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 283340]

Supplier: R&D Systems
FAB2244P
220017-374EA 548.09 USD
220017-374
Anti-CEACAM1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 283340]
Antibodies
Type: Primary
Antigen: CEACAM1
Clonality: Monoclonal
Clone: 283340
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR