Anti-ITGB2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 212701]

Supplier: R&D Systems
FAB1730P
220017-181EA 461.32 USD
220017-181
Anti-ITGB2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 212701]
Antibodies
Type: Primary
Antigen: ITGB2
Clonality: Monoclonal
Clone: 212701
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR