Anti-TREM2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 237920]

Supplier: R&D Systems
FAB17291A
220017-177EA 721.63 USD
220017-177
Anti-TREM2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 237920]
Antibodies
Type: Primary
Antigen: TREM2
Clonality: Monoclonal
Clone: 237920
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR