Anti-IgM Goat Polyclonal Antibody (Fluorescein)

Supplier: R&D Systems
F0118
220016-883EA 273.32 USD
220016-883
Anti-IgM Goat Polyclonal Antibody (Fluorescein)
Antibodies
Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: Fluorescein
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR