Anti-GDF5 Rat Monoclonal Antibody (Biotin) [clone: 90509]

Supplier: R&D Systems
BAM853
220014-662EA 736.09 USD
220014-662
Anti-GDF5 Rat Monoclonal Antibody (Biotin) [clone: 90509]
Antibodies
Type: Primary
Antigen Symbol: GDF5
Antigen Name: growth differentiation factor 5
Antigen Synonyms: BMP-14, Cdmp-1, bp, brp
Clonality: Monoclonal
Clone: 90509
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR