Anti-IL5 Rat Monoclonal Antibody (Biotin) [clone: TRFK4]

Supplier: R&D Systems
BAM705
220014-651EA 793.94 USD
220014-651
Anti-IL5 Rat Monoclonal Antibody (Biotin) [clone: TRFK4]
Antibodies
Type: Primary
Antigen: IL5
Clonality: Monoclonal
Clone: TRFK4
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR