Anti-CLU Mouse Monoclonal Antibody (Biotin) [clone: 350207]

Supplier: R&D Systems
BAM29373
220014-613EA 765.02 USD
220014-613
Anti-CLU Mouse Monoclonal Antibody (Biotin) [clone: 350207]
Antibodies
Type: Primary
Antigen: CLU
Clonality: Monoclonal
Clone: 350207
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR