Anti-IL15 Mouse Monoclonal Antibody (Biotin) [clone: 34593]

Supplier: R&D Systems
BAM247
220014-606EA 837.32 USD
220014-606
Anti-IL15 Mouse Monoclonal Antibody (Biotin) [clone: 34593]
Antibodies
Type: Primary
Antigen: IL15
Clonality: Monoclonal
Clone: 34593
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR