Anti-NOV Rat Monoclonal Antibody (Biotin) [clone: 231210]

Supplier: R&D Systems
BAM19761
220014-589EA 779.48 USD
220014-589
Anti-NOV Rat Monoclonal Antibody (Biotin) [clone: 231210]
Antibodies
Type: Primary
Antigen: NOV
Clonality: Monoclonal
Clone: 231210
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR