Anti-IL17B Mouse Monoclonal Antibody (Biotin) [clone: 174106]

Supplier: R&D Systems
BAM12481
220014-550EA 793.94 USD
220014-550
Anti-IL17B Mouse Monoclonal Antibody (Biotin) [clone: 174106]
Antibodies
Type: Primary
Antigen: IL17B
Clonality: Monoclonal
Clone: 174106
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR