Anti-CDH1 Goat Polyclonal Antibody (Biotin)

Supplier: R&D Systems
BAF748
220014-374EA 851.78 USD
220014-374
Anti-CDH1 Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: CDH1
Clonality: Polyclonal
Clone: 0
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR