Anti-CD80 Goat Polyclonal Antibody (Biotin)

Supplier: R&D Systems
BAF740
220014-367EA 721.63 USD
220014-367
Anti-CD80 Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: CD80
Clonality: Polyclonal
Clone: 0
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR