Anti-TNFRSF10B Goat Polyclonal Antibody (Biotin)

Supplier: R&D Systems
BAF631
220014-298EA 721.63 USD
220014-298
Anti-TNFRSF10B Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: TNFRSF10B
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR