Anti-SIRT5 Sheep Polyclonal Antibody (Biotin)

Supplier: R&D Systems
BAF5914
220014-270EA 736.09 USD
220014-270
Anti-SIRT5 Sheep Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: SIRT5
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR