Anti-GFRAL Sheep Polyclonal Antibody (Biotin)

Supplier: R&D Systems
BAF5728
220014-251EA 808.4 USD
220014-251
Anti-GFRAL Sheep Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen Symbol: GFRAL (Isoform A)
Antigen Name: GDNF family receptor alpha like
Antigen Synonyms: AY457637, Gral
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR