Anti-CDSN Sheep Polyclonal Antibody (Biotin)

Supplier: R&D Systems
BAF5725
220014-250EA 736.09 USD
220014-250
Anti-CDSN Sheep Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: CDSN
Clonality: Polyclonal
Clone: 0
Conjugation: Biotin
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR