Anti-CNTF Goat Polyclonal Antibody (Biotin)

Supplier: R&D Systems
BAF557
220014-231EA 736.09 USD
220014-231
Anti-CNTF Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: CNTF
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR