Anti-SEMA4B Goat Polyclonal Antibody (Biotin)

Supplier: R&D Systems
BAF5485
220014-218EA 736.09 USD
220014-218
Anti-SEMA4B Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: SEMA4B
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR