Anti-PLAUR Goat Polyclonal Antibody (Biotin)

Supplier: R&D Systems
BAF534
220014-196EA 750.55 USD
220014-196
Anti-PLAUR Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: PLAUR
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR