Anti-RAGE Sheep Polyclonal Antibody (Biotin)

Supplier: R&D Systems
BAF4750
220014-078EA 721.63 USD
220014-078
Anti-RAGE Sheep Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: RAGE
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR