Anti-FLT1 Goat Polyclonal Antibody (Biotin)

Supplier: R&D Systems
BAF471
220014-070EA 765.02 USD
220014-070
Anti-FLT1 Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen Symbol: FLT1
Antigen Name: FMS-like tyrosine kinase 1
Antigen Synonyms: AI323757, Flt-1, VEGFR-1, VEGFR1, sFlt1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR