Anti-GLI3 Goat Polyclonal Antibody (Biotin)

Supplier: R&D Systems
BAF3690
220013-864EA 736.09 USD
220013-864
Anti-GLI3 Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen Symbol: GLI3
Antigen Name: GLI family zinc finger 3
Antigen Synonyms: ACLS, GCPS, GLI3-190, GLI3FL, PAP-A, PAPA, PAPA1, PAPB, PHS, PPDIV
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR