Anti-SDC4 Goat Polyclonal Antibody (Biotin)

Supplier: R&D Systems
BAF2918
220013-655EA 765.02 USD
220013-655
Anti-SDC4 Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: SDC4
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR