Anti-CCL8 Goat Polyclonal Antibody (Biotin)

Supplier: R&D Systems
BAF281
220013-627EA 793.94 USD
220013-627
Anti-CCL8 Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: CCL8
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR