Anti-CD300LB Goat Polyclonal Antibody (Biotin)

Supplier: R&D Systems
BAF2580
220013-540EA 736.09 USD
220013-540
Anti-CD300LB Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: CD300LB
Clonality: Polyclonal
Clone: 0
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR