Anti-FGF7 Goat Polyclonal Antibody (Biotin)

Supplier: R&D Systems
BAF251
220013-499EA 765.02 USD
220013-499
Anti-FGF7 Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen Symbol: FGF7
Antigen Name: fibroblast growth factor 7
Antigen Synonyms: HBGF-7, KGF
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR