Anti-TREML1 Goat Polyclonal Antibody (Biotin)

Supplier: R&D Systems
BAF2424
220013-458EA 765.02 USD
220013-458
Anti-TREML1 Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: TREML1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR