Anti-FOSB Goat Polyclonal Antibody (Biotin)

Supplier: R&D Systems
BAF2214
220013-355EA 736.09 USD
220013-355
Anti-FOSB Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen Symbol: FOSB
Antigen Name: FBJ murine osteosarcoma viral oncogene homolog B
Antigen Synonyms: AP-1, G0S3, GOS3, GOSB
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR