Anti-PDGFB Goat Polyclonal Antibody (Biotin)

Supplier: R&D Systems
BAF220
220013-349EA 793.94 USD
220013-349
Anti-PDGFB Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: PDGFB
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR