Anti-CSF3 Goat Polyclonal Antibody (Biotin)

Supplier: R&D Systems
BAF214
220013-325EA 822.86 USD
220013-325
Anti-CSF3 Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: CSF3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR