Anti-IL27RA Goat Polyclonal Antibody (Biotin)

Supplier: R&D Systems
BAF2109
220013-315EA 736.09 USD
220013-315
Anti-IL27RA Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: IL27RA
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR