Anti-HAVCR1 Goat Polyclonal Antibody (Biotin)

Supplier: R&D Systems
BAF1817
220013-177EA 765.02 USD
220013-177
Anti-HAVCR1 Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen Symbol: HAVCR1
Antigen Name: hepatitis A virus cellular receptor 1
Antigen Synonyms: AI503787, KIM-1, TIM-1, Tim1, Timd1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR