Anti-FURIN Goat Polyclonal Antibody (Biotin)

Supplier: R&D Systems
BAF1503
220013-043EA 765.02 USD
220013-043
Anti-FURIN Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen Symbol: FURIN
Antigen Name: furin (paired basic amino acid cleaving enzyme)
Antigen Synonyms: FUR, PACE, PCSK3, SPC1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR