Anti-FSTL3 Goat Polyclonal Antibody (Biotin)

Supplier: R&D Systems
BAF1288
220012-954EA 736.09 USD
220012-954
Anti-FSTL3 Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen Symbol: FSTL3
Antigen Name: follistatin-like 3 (secreted glycoprotein)
Antigen Synonyms: FLRG, FSRP
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR