Anti-FASLG Goat Polyclonal Antibody (Biotin)

Supplier: R&D Systems
BAF126
220012-942EA 793.94 USD
220012-942
Anti-FASLG Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen Symbol: FASLG
Antigen Name: Fas ligand (TNF superfamily, member 6)
Antigen Synonyms: ALPS1B, APT1LG1, APTL, CD178, CD95-L, CD95L, FASL, TNFSF6
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR