Anti-FCGR1A Goat Polyclonal Antibody (Biotin)

Supplier: R&D Systems
BAF1257
220012-940EA 793.94 USD
220012-940
Anti-FCGR1A Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen Symbol: FCGR1A
Antigen Name: Fc fragment of IgG, high affinity Ia, receptor (CD64)
Antigen Synonyms: CD64, CD64A, FCRI, IGFR1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR