Anti-DKK3 Goat Polyclonal Antibody (Biotin)

Supplier: R&D Systems
BAF1118
220012-857EA 779.48 USD
220012-857
Anti-DKK3 Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: DKK3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR