Anti-F11R Goat Polyclonal Antibody (Biotin)

Supplier: R&D Systems
BAF1077
220012-829EA 721.63 USD
220012-829
Anti-F11R Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: F11R
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR