Anti-CTSD Goat Polyclonal Antibody (Biotin)

Supplier: R&D Systems
BAF1014
220012-788EA 793.94 USD
220012-788
Anti-CTSD Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: CTSD
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR