Anti-GMCSF1 Goat Polyclonal Antibody

Supplier: R&D Systems
AF987
220012-735EA 692.71 USD
220012-735
Anti-GMCSF1 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen Symbol: GM-CSF
Antigen Name:
Antigen Synonyms:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Cat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR