Anti-IFNG Goat Polyclonal Antibody

Supplier: R&D Systems
AF985
220012-733EA 678.25 USD
220012-733
Anti-IFNG Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IFNG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR