Anti-FGF19 Goat Polyclonal Antibody

Supplier: R&D Systems
AF969
220012-719EA 678.25 USD
220012-719
Anti-FGF19 Goat Polyclonal Antibody
Antibodies
Blockade of Receptor-ligand Interaction

Type: Primary
Antigen Symbol: FGF19
Antigen Name: fibroblast growth factor 19
Antigen Synonyms: -
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR