Anti-BAD Rabbit Polyclonal Antibody

Supplier: R&D Systems
AF819
220012-603EA 504.71 USD
220012-603
Anti-BAD Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: BAD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR