Anti-GDF7 Goat Polyclonal Antibody

Supplier: R&D Systems
AF779
220012-566EA 649.32 USD
220012-566
Anti-GDF7 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen Symbol: GDF7
Antigen Name: growth differentiation factor 7
Antigen Synonyms: BMP12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR