Anti-CSF2RA Goat Polyclonal Antibody

Supplier: R&D Systems
AF706
220012-511EA 663.78 USD
220012-511
Anti-CSF2RA Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CSF2RA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR