Anti-BIRC5 Goat Polyclonal Antibody

Supplier: R&D Systems
AF6471
220012-475EA 707.17 USD
220012-475
Anti-BIRC5 Goat Polyclonal Antibody
Antibodies
Simple Western

Type: Primary
Antigen: BIRC5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR